Learn More
Medchemexpress LLC Cyclo[Arg-Gly-Asp-D-Phe-Lys(Azide)] | 2767993-86-0 | 99.6% | 700.75 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1030310MG
Encompass_Preferred
Cyclo[Arg-Gly-Asp-D-Phe-Lys(Azide)] is a derivative of Cyclo(-RGDfK) (HY-P0023). This compound is a click chemistry reagent containing an azide group. Click chemistry is widely used in research due to its high yield, high specificity, and simplicity, with significant potential for binding various molecules such as nucleic acids, lipids, and proteins.
  • Derivative of Cyclo(-RGDfK)
  • Contains an azide group
  • Functions as a click chemistry reagent
  • Exhibits high yield
  • Offers high specificity
  • Provides simplicity in binding various molecules
  • Binds nucleic acids, lipids, and proteins
  • Applicable in various research fields
Catalog No. 50-004-25143