Learn More
Medchemexpress LLC Cyclo[Arg-Gly-Asp-D-Phe-Lys(Azide)] (Cyclo[RGDfK(Azide)]) | 2767993-86-0 | 99.56% | 700.75 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP103035MG
Encompass_Preferred
Cyclo[Arg-Gly-Asp-D-Phe-Lys(Azide)] is a derivative of Cyclo(-RGDfK). This product is a click chemistry reagent that contains an azide group. Click chemistry is a valuable tool for binding between nucleic acids, lipids, proteins, and other molecules, and is favored for its high yield, high specificity, and simplicity.
  • Derivative of Cyclo(-RGDfK)
  • Click chemistry reagent
  • Contains an azide group
  • High yield
  • High specificity
  • Simple to use
  • Useful for binding nucleic acids, lipids, and proteins
  • White to off-white solid appearance
Catalog No. 50-004-25147