Learn More
Medchemexpress LLC Cyclo(Arg-Gly-Asp-D-Tyr-ε-azido-Nle) | 1392269-21-4 | 99.2% | 645.67 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP50255MG
Encompass_Preferred
Cyclo(Arg-Gly-Asp-D-Tyr-ε-azido-Nle), also known as Azido-c(RGDyK), is a polypeptide composed of arginine, glycine, aspartic acid, and tyrosine. This compound is primarily utilized in the synthesis of [18F]FPyKYNE-c(RGDyK). It is intended for research use only.
  • Polypeptide composed of arginine, glycine, aspartic acid, and tyrosine
  • Used for synthesis of [18F]FPyKYNE-c(RGDyK)
  • Intended for research use only
Catalog No. 50-003-98613