Learn More
Carbosynth LLC. Cyclo(Arg-Gly-Asp-D-Tyr-Lys), 1mg, MW: (M.W. 619.68), Molecular formula: C27H41N9O8
SDP
Supplier:  Carbosynth LLC. PCI3662PI1MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: cyclo (Arg-Gly-Asp-D-Tyr-Lys), c(RGDyK), Application: alphavbeta3 Integrin Binding RGD Peptide RGD Tumor Targeting and Tumor Imaging Peptide (Requires further derivatization before use), Storage Temperature: -20C, References: R. Haubner, H.J. Wester, F. Burkhart, R. Senekowitsch-Schmidtky, W. Weber, S.L. Goodman, H. Kessler, and M. Schwaiger, J. NuCLEAR™ Med., 42, 326 (2001), X. Chen, R. Park. A.H. Shahinian, M. Tohme, V. Khankaldyyan, M.H. Bozorgzdeh, J.R. Bading, R. Moats, W.E. Laug, and P.S. Conti, Nucl. Med. Biol., 31, 179 (2004)., X. Chen, P.S. Conti, and R.A. Moats, Cancer Res., 641, 8009 (2004).
Catalog No. 50-168-6276