Learn More
Carbosynth LLC. Cyclo(Arg-Gly-Asp-D-Tyr-Lys), 25mg, MW: (M.W. 619.68), Molecular formula: C27H41N9O8
Supplier: Carbosynth LLC. PCI3662PI25MG
Synonyms: cyclo (Arg-Gly-Asp-D-Tyr-Lys), c(RGDyK), Application: alphavbeta3 Integrin Binding RGD Peptide RGD Tumor Targeting and Tumor Imaging Peptide (Requires further derivatization before use), Storage Temperature: -20°C, References: R. Haubner, H.J. Wester, F. Burkhart, R. Senekowitsch-Schmidtky, W. Weber, S.L. Goodman, H. Kessler, and M. Schwaiger, J. NuCLEAR™ Med., 42, 326 (2001), X. Chen, R. Park. A.H. Shahinian, M. Tohme, V. Khankaldyyan, M.H. Bozorgzdeh, J.R. Bading, R. Moats, W.E. Laug, and P.S. Conti, Nucl. Med. Biol., 31, 179 (2004)., X. Chen, P.S. Conti, and R.A. Moats, Cancer Res., 641, 8009 (2004).