Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclo(CKLIIF) (TFA) | 98.1% | 831.99 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYP10221A5MG

Encompass_Preferred

Cyclo(CKLIIF) TFA is a dual inhibitor for hypoxia inducible factor (HIF) 1 and 2. It functions by disrupting the interaction of both HIF1-α and HIF2-α with HIF1-β, exhibiting an affinity (KD) for HIF1-α and HIF2-α PAS-B domains of 2.6 μM and 2.2 μM, respectively. This product is suitable for use as a laboratory chemical in the manufacture of substances.

  • Dual inhibitor for HIF 1 and 2
  • Disrupts interaction of HIF1-α and HIF2-α with HIF1-β
  • Affinity for HIF1-α PAS-B domain is 2.6 μM
  • Affinity for HIF2-α PAS-B domain is 2.2 μM
  • Molecular weight: 831.99
  • Appearance: White to off-white solid
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-24424


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.