Learn More
Medchemexpress LLC Cyclo(Gly-Gln) | 52662-00-7 | 98.1% | 50 MG

Supplier: Medchemexpress LLC HY13111150MG
Cyclo(Gly-Gln) is a cyclic dipeptide.
- Molecular weight of 185.18
- Chemical formula C7H11N3O3
- Appears as a solid
- Color ranges from white to off-white
- Sequence is Cyclo(Gly-Gln)
- Shortened sequence is Cyclo(GQ)
- Solubility in H2O is 13.89 mg/mL (75.01 mM)
- Powder can be stored at -80°C for 2 years or -20°C for 1 year
- Solution can be stored at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.