Learn More
Medchemexpress LLC Cyclo(Gly-L-Pro) | 3705-27-9 | 99.86% | 154.17 | 250 MG

Supplier: Medchemexpress LLC 250MGHYP1936
Cyclo(Gly-L-Pro) is an Alkaloids product that can be isolated from the roots of Panax notoginseng. It acts as an inhibitor of LPS-induced TNF-alpha production in mouse RAW264.7 cells with an IC50 of 4.5 μg/mL. This product is intended for research use only.
- High purity: 99.86%
- Soluble in water and DMSO
- Recommended storage conditions for powder: -80°C for 2 years; -20°C for 1 year
- Recommended storage conditions in solvent: -80°C for 6 months; -20°C for 1 month
- Store under nitrogen, away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.