Learn More
Medchemexpress LLC Cyclo(Gly-L-Pro) | 3705-27-9 | 99.9% | 154.17 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP193650MG
Encompass_Preferred
Cyclo(Gly-L-Pro) is an alkaloids product isolated from the roots of Panax notoginseng (Burk.)F.H.Chen. It inhibits LPS-induced TNF-alpha production in mouse RAW264.7 cells. This product is for research use only.
  • Inhibits LPS-induced TNF-alpha production in mouse RAW264.7 cells (IC50 4.5 μg/mL)
Catalog No. 50-004-23515