Learn More
Medchemexpress LLC Cyclo(His-Phe) TFA | 147600-13-3 | 99.9% | 398.34 | 5 MG

Supplier: Medchemexpress LLC HYP5028A5MG
Cyclo(His-Phe) TFA is a high-purity cyclic dipeptide compound with a molecular weight of 398.34. It is supplied as a white to off-white solid, offering reliable performance for various research and laboratory applications.
- Purity of 99.9%
- Supplied as a white to off-white solid
- For powder, store at -80°C for 2 years or -20°C for 1 year
- For solutions, store at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.