Learn More
Medchemexpress LLC Cyclo(L-Phe-trans-4-OH-L-Pro) | 118477-06-8 | 260.29 | 5 MG

Supplier: Medchemexpress LLC HYN125135MG
Cyclo(L-Phe-trans-4-OH-L-Pro) is an antifungal peptide.
- Purity: 99.76%
- Appearance: Solid
- Color: White to off-white
- Target: Fungal
- Pathway: Anti-infection
- Initial source: Microorganisms, specifically Lactobacillus plantarum
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.