Learn More
Cayman Chemical Cyclo L-Pro-L-Val 25mg
SDP
Supplier:  Cayman Chemical 2796125 MG
Encompass_Preferred
A diketopiperazine; active against S. aureus and B. subtilis (MICs = 16.3 and 18.2 UG/ml, respectively); modulates LuxR-dependent quorum sensing systems but not lacZ-based reporter fusions; inhibits activation of a LuxR-dependent E. coli biosensor by 3-oxo-hexanoyl-homoserine lactone (IC50 = 0.4 mM) and activates violacein pigment production in the LuxR-dependent C. violaceum acyl homoserine lactone reporter strain CV026
Catalog No. 50-294-0866