Learn More
Medchemexpress LLC Cyclo(L-Pro-L-Val) | 2854-40-2 | 98.1% | 196.25 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP1938
Encompass_Preferred
Cyclo(L-Pro-L-Val) is an antimicrobial and anti-inflammatory agent that exhibits toxic activity against plant pathogens like R. fascians LMG 3605, with potency comparable to Chloramphenicol. It can inhibit the phosphorylation of IKKα, IKKβ, NF-κB, and the activation of iNOS and COX-2, thereby demonstrating anti-inflammatory activity. This compound is suitable for research in biopesticides for agriculture and inflammation-related diseases.
  • Antimicrobial agent
  • Anti-inflammatory agent
  • Toxic activity against plant pathogens such as R. fascians LMG 3605
  • Potency may be comparable to Chloramphenicol
  • Inhibits phosphorylation of IKKα, IKKβ, NF-κB
  • Inhibits activation of iNOS and COX-2
  • Can be used in biopesticide research in agriculture
  • Can be used in inflammation-related disease research
Catalog No. 50-004-26921