Learn More
Cayman Chemical Cyclo L-Trp-L-Trp 500mg
SDP
Supplier:  Cayman Chemical 23494500 MG
Encompass_Preferred
A cyclic dipeptide; used as a substrate for indole prenyltransferases in the synthesis of mono- and diprenylated indolines; inhibits growth of 41 out of 49 strains of multidrug resistant A. baumannii (MICs = 12.5-25 UG/ml) as well as B. subtilis, M. luteus, S. aureus, S. cerevisiae, A. niger, and C. albicans (MICs = 12.5-50 UG/ml)
Catalog No. 50-299-4688