Learn More
Medchemexpress LLC Cyclo(Phe-Gly) | 5037-75-2 | MFCD00190926 | 98.5% | 204.23 g·mol^-1 | C11H12N2O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW29587310MM/1ML
Encompass_Preferred
Cyclo(Phe-Gly) is a cyclic dipeptide reported to exhibit antimicrobial and anticancer activities, originally isolated from Streptomyces YIM 64018. The compound is offered as a high-purity solid and as a solution formulation for use in biological assays and natural product research.
  • Cyclic dipeptide with reported antimicrobial and anticancer activity.
  • Provided as powder and as a 10 mM solution in DMSO for convenience in assays.
  • High purity suitable for biological research.
  • Commonly used in cellular assays and natural product studies.
  • Molecular formula C11H12N2O2; molecular weight 204.23 g·mol⁻1.
Catalog No. 50-002-82529