Learn More
Medchemexpress LLC Cyclo(Phe-Gly) | 5037-75-2 | MFCD00190926 | 98.5% | 204.23 g/mol | C11H12N2O2 | 5 MG

Supplier: Medchemexpress LLC HYW2958735MG
Cyclo(Phe-Gly) is a cyclic dipeptide used in research for its reported antimicrobial and anticancer activities. It was isolated from broth culture of endophytic Streptomyces YIM 64018 associated with Paraboea sinensis, and is supplied as a solid and as a DMSO solution for laboratory use.
- Cyclic dipeptide with reported antimicrobial and anticancer activity.
- CAS number 5037-75-2 for unambiguous chemical identification.
- Molecular formula C11H12N2O2 and molecular weight 204.23 g/mol.
- High purity (98.5%) suitable for research applications.
- Available as small solid quantities and 10 mM DMSO solutions.
- Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.