Learn More
Medchemexpress LLC Cyclo(RADfK) | 756500-23-9 | 96.4% | 617.70 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP003110MG
Encompass_Preferred
Cyclo(RADfK) is a negative control for cyclo(-RGDfK), a RGD peptide known to modulate cell adhesion and interact with integrin family members. It can be used as a control for studies involving Cyclo(-RGDfK).
  • Serves as a negative control for RGD peptides
  • Modulates cell adhesion properties
  • Recognized by integrin family members
  • Provided as a white to off-white solid
  • Soluble in H2O and PBS (100 mg/mL), requiring sonication
Catalog No. 50-004-26493