Learn More
Medchemexpress LLC Cyclo(RADfK) | 756500-23-9 | >96.4% | 617.70 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP0031
Encompass_Preferred
Cyclo(RADfK) serves as a negative control for Cyclo(-RGDfK). It is useful in studies involving RGD peptides, which are known modulators of cell adhesion and are recognized by various members of the integrin family.
  • Functions as a negative control for RGD peptides
  • Modulates cell adhesion
  • Recognized by integrin family members
  • Soluble in water and PBS at 100 mg/mL
  • Targets integrin
  • Involved in cytoskeleton pathway
Catalog No. 50-004-26751