Learn More
CPC Scientific Cyclo(-Arg-Ala-Asp-D-Phe-Lys-) (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific RGDP003B
Encompass
This peptide is a negative control for the cyclo (-RGDfK-), the RGD peptide. RGD peptides are modulators of cell adhesion and are recognized by several members of the integrin family. This peptide has low affinity binding to integrin peptides.SEQUENCE: Cyclo(-Arg-Ala-Asp-D-Phe-Lys-) (trifluoroacetate salt)ONE-LETTER SEQUENCE: Cyclo(-RADfK-)MOLECULAR FORMULA: C28H43N9O7MOLECULAR WEIGHT: 617.70STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [756500-23-9]SYNONYMS: Cyclo(-Arg-Ala-Asp-D-Phe-Lys), c(RADfK)RESEARCH AREA: CancerREFERENCES:Wester, H. et al. J. Nucl. Med. 46, 1940 (2005)Janssen, M. et al. Canc. Res. 62, 6146 (2002)SKU(s): RGDP-003A, RGDP-003B
Catalog No. 50-211-0377