Learn More
CPC Scientific Cyclo(-Arg-Ala-Asp-D-Phe-Glu-) (ammonium salt) 5MG
SDP
Supplier:  CPC Scientific RGDP002C
Encompass
This peptide is a negative control for the cyclo (-RGDfE) peptide. RGD peptides are modulators of cell adhesion, and are recognized by several members of the integrin family. This peptide has a low affinity binding to integrin peptides.SEQUENCE: Cyclo(-Arg-Ala-Asp-D-Phe-Glu-) (ammonium salt)ONE-LETTER SEQUENCE: Cyclo(-RADfE-)MOLECULAR FORMULA: MOLECULAR WEIGHT: 618.65STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [756500-23-9]SYNONYMS: RESEARCH AREA: CancerCATEGORY: Fibronectin and RGD Sequences »REFERENCES:Wester, H. et al. J. Nucl. Med. 46, 1940 (2005)Janssen, M. et al. Canc. Res. 62, 6146 (2002)SKU(s): RGDP-002A, RGDP-002B, RGDP-002C
Catalog No. 50-211-0375