Learn More
Apexbio Technology LLC Cyclo (-RGDfC), 25mg CAS# 862772-11-0
SDP
Supplier:  Apexbio Technology LLC A879025
Encompass_Preferred
RGD tumor targeting peptide can bind to v3 receptor. c (RGDfC) can be easily conjugated to convistatin or drug surface. Cyclo (-RGDfC) is an integrin avb3-affine polypeptide, a circular RGDfc sequence. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8401