Learn More
Medchemexpress LLC Cyclo(-RGDfK) | 161552-03-0 | 99.8% | 603.67 | 25 MG

Supplier: Medchemexpress LLC 25MGHYP0023
Cyclo(-RGDfK) is a potent and selective inhibitor of the αvβ3 integrin, with an IC50 of 0.94 nM. It targets tumor microvasculature and cancer cells by specifically binding to the αvβ3 integrin on the cell surface.
- Potent and selective αvβ3 integrin inhibitor.
- IC50 of 0.94 nM.
- Targets tumor microvasculature and cancer cells.
- Appearance: solid, white to off-white.
- Chemical sequence: Cyclo(Arg-Gly-Asp-{dPhe}-Lys).
- Shortened sequence: Cyclo(RGD-{dPhe}-K).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.