Learn More
Medchemexpress LLC Cyclo(-RGDfK) TFA | 500577-51-5 | 99.6% | 717.69 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP0023A5MG
Encompass_Preferred
Cyclo(-RGDfK) TFA is a potent and selective inhibitor of the αvβ3 integrin, demonstrating an IC50 of 0.94 nM. This compound effectively targets tumor microvasculature and cancer cells through specific binding to the αvβ3 integrin on the cell surface. This product is for research use only, and can be used in microPET imaging studies.
  • Potent and selective inhibitor of αvβ3 integrin with an IC50 of 0.94 nM.
  • Effectively targets tumor microvasculature and cancer cells.
  • Binds specifically to the αvβ3 integrin on the cell surface.
  • Suitable for research use only.
  • Can be used in microPET imaging applications.
Catalog No. 50-004-26150