Learn More
Medchemexpress LLC Cyclo(RGDyC) | 1206475-79-7 | 98.3% | 594.64 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP584010MG
Encompass_Preferred
Cyclo(RGDyC) is a cyclic pentapeptide known for its anti-angiogenic properties. It demonstrates the ability to inhibit the formation of new blood vessels, making it a subject of interest in various research areas.
  • Cyclic pentapeptide
  • Exhibits anti-angiogenic abilities
  • Can be combined with liposome delivery systems
  • Useful for research on ocular neovascular diseases
  • Applicable in cancer research
Catalog No. 50-003-99165