Learn More
Cayman Chemical cyclo RGDyCtrIfluoroACTAt 10mg
SDP
Supplier:  Cayman Chemical 3494510 MG
Encompass_Preferred
A cyclic pentapeptide; has been used to generate doxorubicin-containing liposomes and fluorescent silicon nanoparticles to target tumors and ocular neovasculature, respectively
Catalog No. 50-291-0938