Learn More
Apexbio Technology LLC Cyclo(RGDyK) 250612-42-1 25mg
SDP
Supplier:  Apexbio Technology LLC B601525
Encompass_Preferred
Cyclo RGDyK (CAS 250612-42-1) is a cyclic pentapeptide antagonist targeting integrin V 3 It is designed to competitively inhibit the interaction between integrin V 3 and ligands containing RGD motifs thereby modulating integrin-mediated cellular processes Cyclo RGDyK exerts its biological activity primarily through antagonism of V 3 integrin In preclinical experiments Cyclo RGDyK demonstrates selective inhibition of V 3 integrin with an IC50 of approximately 20 nM Based on these pharmacological properties Cyclo RGDyK holds research potential in integrin-mediated cellular process studies and tumor targeting applications
Catalog No. 50-000-92410