Learn More
Medchemexpress LLC Cyclo(rgdyk) (trifluoroacetate) | 250612-42-1 | 99.9% | 847.72 g/mol | C31H43F6N9O12 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10056310MG
Encompass_Preferred
Cyclo(RGDyK) trifluoroacetate is a cyclic pentapeptide that potently and selectively inhibits the αVβ3 integrin (IC50 = 20 nM). Supplied as the trifluoroacetate salt, it is intended for research use in studies of integrin-mediated cell adhesion, migration, and angiogenesis.
  • Potent αVβ3 inhibition (IC50 20 nM).
  • High purity suitable for research (99.9%).
  • Trifluoroacetate salt form to aid handling and solubility.
  • Well characterized molecular weight (847.72 g/mol).
  • Useful for assays of cell adhesion, migration, and angiogenesis.
Catalog No. 50-002-11932