Learn More
Medchemexpress LLC Cyclo(RLsKDK) TFA | 99.66% | 841.88 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP1676A5MG
Encompass_Preferred
Cyclo(RLsKDK) TFA is a specific metalloproteinase ADAM8 inhibitor (IC50 182 nM) with potential applications in inflammatory diseases and cancer. It promotes ADAM8 activation and CD23 shedding in vitro, and significantly reduces tumor load and improves survival in pancreatic ductal adenocarcinoma (PDAC) mouse models.
  • Specific inhibitor of metalloproteinase ADAM8
  • Applications in inflammatory diseases and cancer
  • Promotes ADAM8 activation and CD23 shedding
  • Increases activity of pro-ADAM8
  • Promotes growth of Panc1_ctrl and Panc1_A8 cells
  • Causes ERK1/2 phosphorylation
  • Reduces tumor load in mouse models
  • Improves survival in PDAC mice
  • Reduces soluble ADAM8 (sADAM8) content
  • Decreases pERK1/2 activation and PDAC metastasis
Catalog No. 50-004-23668