Learn More
Medchemexpress LLC Cycloartenol | 469-38-5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN72551MG
Encompass_Preferred
Cycloartenol is a phytosterol compound and a key precursor for the biosynthesis of numerous sterol compounds. It exhibits various pharmacological activities including anti-inflammatory, anti-tumor, antioxidant, antibiosis, and anti-Alzheimer's disease effects. It also plays a significant role in plant growth and development.
  • Inhibits migration of glioma cells.
  • Suppresses phosphorylation of p38 MAP kinase.
  • Induces Sub-G1 cell cycle arrest and apoptosis in glioma U87 cells.
  • Significantly alters the expression of Bax and Bcl-2.
  • Acts as a precursor for sterol biosynthesis.
Catalog No. 50-004-17235