Learn More
Cayman Chemical Cycloastragenol 25mg
SDP
Supplier:  Cayman Chemical 1172225 MG
Encompass_Preferred
A telomerase activator that can slow telomere loss, increase replicative capacity, and improve immune function in CD8+ T lymphocytes from HIV-1-infected patients
Catalog No. 50-295-2482