Learn More
Cayman Chemical CyclobenzaprIn N-oxIde 25mg
SDP
Supplier:  Cayman Chemical 3669125 MG
Encompass_Preferred
A metabolite of cyclobenzaprine; a potential impurity in commercial preparations of cyclobenzaprine
Catalog No. 50-290-9450