Learn More
Medchemexpress LLC Cyclobenzaprine hydrochloride | 6202-23-9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0740R100MG
Encompass_Preferred
Cyclobenzaprine (MK130) hydrochloride is an orally active 5-HT2 receptor antagonist that acts centrally to provide skeletal muscle relaxation, alleviate muscle spasms, and reduce pain. It also possesses antiparasitic activity. Cyclobenzaprine hydrochloride shows promise for research in acute, painful skeletal muscle disorders and infectious diseases.
  • Acts centrally to provide skeletal muscle relaxation
  • Alleviates muscle spasms and reduces pain
  • Possesses antiparasitic activity
  • Shows promise for research in acute, painful skeletal muscle disorders
  • Shows promise for research in infectious diseases
Catalog No. 50-004-06750