Learn More
Apexbio Technology LLC β-Cyclodextrin 7585-39-9 500mg
SDP
Supplier:  Apexbio Technology LLC C6523500
Encompass_Preferred
beta-Cyclodextrin (CAS 7585-39-9) is a cyclic oligosaccharide composed of -D-glucopyranoside units linked by -(1 4) glycosidic bonds beta-Cyclodextrin enhances the aqueous solubility of hydrophobic active molecules through molecular encapsulation beta-Cyclodextrin exerts its biological activity primarily by encapsulating hydrophobic compounds improving their bioavailability In cell-based viral replication assays beta-Cyclodextrin demonstrates antiviral activity with IC50 values ranging from 3 5 to 7 5 mM against H1N1 influenza virus strains depending on experimental conditions Based on these pharmacological properties beta-Cyclodextrin holds research potential in pharmaceutical formulation and antiviral screening
Catalog No. 50-000-90687