Learn More
Sigma Aldrich Fine Chemicals Biosciences Cyclohexane ACS reagent 991L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 1791911L
Encompass
Cyclohexane is a cyclic alkane that predominantly exists in chair conformation due to higher stability. It participates as starting reagent in the photonitrosylation process (PNC process) and affordsµ-caprolactum. Its oxidation by employing various oxidants such as hydrogen peroxide tert-butyl hydroperoxide and molecular oxygen has been reported.
Catalog No. 50-289-7329