Learn More
Medchemexpress LLC Cyclohexaneacetic acid | 5292-21-7 | MFCD00001518 | 99.8% | 142.20 g·mol⁻¹ | C8H14O2 | 1 ML

Supplier: Medchemexpress LLC HYW01865310MM/1ML
Cyclohexaneacetic acid is supplied as a ready-to-use 10 mM solution in DMSO for research use. The product is characterized by high purity and is accompanied by certificate of analysis and safety documentation; storage recommendations and stability data are provided for both powder and solution forms.
- Ready-to-use 10 mM solution in DMSO.
- High purity (99.78%).
- Molecular formula C8H14O2 and molecular weight 142.20 g·mol⁻¹.
- Supplied with COA and SDS for quality and safety review.
- Storage guidance provided for powder and in-solution stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.