Learn More
Medchemexpress LLC Cyclohexanecarboxylic acid, 1-phenyl-, 2-(4-morpholinyl)ethyl ester, hydrochloride (1:1) | 75136-54-8 | 99.8% | 353.88 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHY18100A
Encompass_Preferred
PRE-084 hydrochloride is a highly selective σ1 receptor (S1R) agonist, with an IC50 of 44 nM. This product is intended for research and analytical applications.
  • Highly selective σ1 receptor (S1R) agonist with an IC50 of 44 nM.
  • Exhibits good neuroprotective effects.
  • Can improve motor function and motor neuron survival in mice.
  • Can ameliorate myocardial ischemia-reperfusion injury in rats by activating the Akt-eNOS pathway.
Catalog No. 50-003-65143