Learn More
Medchemexpress LLC Cyclohexanepropanoic acid, 2-(dimethylamino)ethyl ester | 15585-86-1 | MFCD00868086 | 98.0% | 227.34 g/mol | C13H25NO2 | 50 MG

Supplier: Medchemexpress LLC HY11863550MG
Cyprodenate is an orally active, blood-brain barrier-penetrant stimulant supplied for laboratory research use only. The product is provided as a 50 mg vial at high purity and is suitable for in vitro and in vivo formulation following laboratory protocols.
- Has CAS number 15585-86-1.
- Molecular formula C13H25NO2 and molecular weight 227.34 g/mol.
- Purity 98.0% by manufacturer specification.
- Highly soluble in DMSO (≥ 200 mg/mL) and formulatable for in vivo use (≥ 5 mg/mL in recommended vehicles).
- Appearance: off-white to light brown liquid; density ~0.958 g/cm3.
- Storage: store product at 4°C and protect from light; in solution store at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.