Learn More
Medchemexpress LLC Cyclohexene, 1-methyl- | 591-49-1 | 96.17 | 50 G

Supplier: Medchemexpress LLC HYW00433850G
1-Methylcyclohexene is a male insect sex pheromone with a purity of 96.58%. This product is suitable for various research applications. It is supplied as a colorless to light yellow liquid with a molecular weight of 96.17 and a chemical formula of C7H12. Its CAS number is 591-49-1.
- Categorized as a male insect sex pheromone
- Purity of 96.58%
- Appears as a colorless to light yellow liquid
- Molecular formula C7H12
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.