Learn More
Enzo Life Sciences Cycloheximide (5g). CAS: 66-81-9
SDP
Supplier:  Enzo Life Sciences ALX380269G005
Encompass_Preferred
Antibiotic. Inhibits protein synthesis in eukaryotes by interfering with the translocation step. Induces apoptosis in tumor cell lines but can also delay or inhibit apoptosis induced by other agents. Widely used for selection of resistant strains of yeast and fungi, controlled inhibition of protein synthesis for detection of short-lived proteins and super-induction of protein expression. At 10µg/ml cycloheximide totally inhibits protein synthesis in mouse thymocytes. Alternative name: Actidione, Naramycin A, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide. Purity: ≥90% (HPLC). Appearance: White to light yellow powder. Solubility: Soluble in acetone, chloroform, ethanol, and methanol. Nearly insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9000