Learn More
Cayman Chemical Cyclohexnoyl-Coenzyme A 1mg
SDP
Supplier:  Cayman Chemical 235041 MG
Encompass_Preferred
An acyl-CoA that contains a cyclohexane group; formed through conversion of CHC by a succinyl-CoA transferase, and degraded by a dehydrogenase; converted to hippuric acid in submitochondrial fractions isolated from guinea pig liver
Catalog No. 50-299-5347