Learn More
Medchemexpress LLC Cyclohexyldiphenylphosphine | 6372-42-5 | 100.0% | 268.34 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW013327100G
Encompass_Preferred
Cyclohexyldiphenylphosphine is a chemical compound commonly used as a ligand in coordination chemistry and organometallic synthesis. It facilitates various catalytic reactions by acting as a stabilizing agent. Additionally, Cyclohexyldiphenylphosphine serves as a ligand for some nickel(II) complexes and has demonstrated anti-cancer properties against SNO cancer cells when complexed with silver.
  • Used as a ligand in coordination chemistry
  • Employed in organometallic synthesis
  • Acts as a stabilizing agent in catalytic reactions
  • Ligand for certain nickel(II) complexes
  • Exhibits anti-cancer properties against SNO cancer cells in silver complexes
Catalog No. 50-004-54283