Learn More
Medchemexpress LLC Cyclohexyldiphenylphosphine | 6372-42-5 | 99.99% | 268.34 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01332750G
Encompass_Preferred
Cyclohexyldiphenylphosphine is a chemical compound commonly used as a ligand in coordination chemistry and organometallic synthesis. It facilitates various catalytic reactions by acting as a stabilizing agent. This compound is a ligand for some nickel(II) complexes and exhibits anti-cancer properties against SNO cancer cells when formed in a complex with silver.
  • Used as a ligand in coordination chemistry and organometallic synthesis.
  • Facilitates various catalytic reactions.
  • Acts as a stabilizing agent.
  • Ligand for some nickel(II) complexes.
  • Exhibits anti-cancer properties against SNO cancer cells when complexed with silver.
Catalog No. 50-004-54626