Learn More
eMolecules​ 931-51-1 | Cyclohexylmagnesium chloride, 1M in THF | Synthonix - Stock | MFCD00003816 | 142.910 | C6H11ClMg | 0.000 | [Cl-].[Mg+]C1CCCCC1 | 50ml | 437237020
Supplier:  eMolecules​ 437237020
Encompass_Preferred
Cyclohexylmagnesium chloride, 1M in THF | Synthonix - Stock | 931-51-1 | MFCD00003816 | 142.910 | C6H11ClMg | 0.000 | [Cl-].[Mg+]C1CCCCC1 | 50ml | 437237020
Catalog No. 50-269-7764