Learn More
Elemental Scientific Inc CYCLONIC SPRAY CHAMBER QUARTZ
SDP
Supplier:  Elemental Scientific Inc ES3160111123
Encompass_Preferred
O-ring free, baffled quartz cyclonic spray chamber for Thermo X-Series ICPMS with 1/4-28 threaded auxiliary gas port.
Catalog No. NC0317423