Learn More
Medchemexpress LLC Cyclooct-4-en-1-yl (2-(2-(2-(2-aminoethoxy)ethoxy)ethoxy)ethyl)carbamate | 1800507-93-0 | 95.0% | C17H32N2O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14117810MG
Encompass_Preferred
TCO-PEG3-amine is a trans-cyclooctene (TCO)-functionalized PEG3 linker terminating in a primary amine, designed for bioconjugation and linker assembly in targeted degrader and chemical biology workflows. It combines a hydrophilic PEG spacer with a fast-reacting TCO handle for bioorthogonal labeling and a free amine for conventional coupling chemistries.
  • Provides a trans-cyclooctene handle for rapid bioorthogonal click reactions.
  • Contains a PEG3 spacer to improve solubility and reduce steric hindrance.
  • Features a terminal primary amine for amide bond formation and conjugation.
  • Suitable for PROTAC linker assembly, bioconjugation, and labeling applications.
  • Offered as a high-purity research reagent with defined molecular properties.
Catalog No. 50-002-58925