Learn More
Medchemexpress LLC Cyclooxygenase 2 antibody (YA2541) | 10 UL

Supplier: Medchemexpress LLC HYP8279610UL
Cyclooxygenase 2 Antibody (YA2541) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Cyclooxygenase 2. This antibody is suitable for use in Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), and Immunoprecipitation (IP) applications. It is supplied in a liquid formulation and is affinity purified.
- Host: Rabbit
- Reactivity: Mouse
- Conjugation: Non-conjugated
- Formulation: 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
- Storage: Stored at -20°C for 1 year
- Appearance: Liquid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.