Learn More
Apexbio Technology LLC Cyclopamine, 10mg. Cas: 4449-51-8 MFCD: MFCD09878269. Hedgehog (Hh) signaling Inhibitor
SDP
Supplier:  Apexbio Technology LLC A834010
Encompass_Preferred
Cyclopamine is a naturally occurring Hedgehog (Hh)?specific small?molecule signaling steroidal alkaloid inhibitor, causes a profound inhibition of tumor growth, has significant anti?invasive, anti?proliferative and anti?estrogenic potency in human breast cancer cells [2] [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5119