Learn More
Strem, An Ascensus Company CAS# 12154-95-9. 50g. Cyclopentadienyliron dicarbonyl dimer, 99%. MFCD00001436
SDP
Supplier:  Strem, An Ascensus Company 26090050G
Encompass
CAS# 12154-95-9. 50g. Cyclopentadienyliron dicarbonyl dimer, 99%. MFCD00001436. Molecular Weight: 353.92. Molecular Formula: [C5H5Fe(CO)2]2. Color/form: red to purple xtl. Strem# 26-0900. http://www.strem.com/catalog/v/26-0900/
Catalog No. 50-901-10993