Learn More
Strem, An Ascensus Company CAS# 12079-65-1. 5g. Cyclopentadienylmanganese tricarbonyl, 98% Cymantrene. MFCD00001440
SDP
Supplier:  Strem, An Ascensus Company 2503905G
Encompass
CAS# 12079-65-1. 5g. Cyclopentadienylmanganese tricarbonyl, 98% Cymantrene. MFCD00001440. Molecular Weight: 204.06. Molecular Formula: C5H5Mn(CO)3. Color/form: yellow xtl. Strem# 25-0390. http://www.strem.com/catalog/v/25-0390/
Catalog No. 50-901-10899