Learn More
Strem, An Ascensus Company CAS# 34767-44-7. 1g. Cyclopentadienylzirconium trichloride, min. 98%. MFCD00015756
SDP
Supplier:  Strem, An Ascensus Company 4011001G
Encompass
CAS# 34767-44-7. 1g. Cyclopentadienylzirconium trichloride, min. 98%. MFCD00015756. Molecular Weight: 262.67. Molecular Formula: (C5H5)ZrCl3. Color/form: off-white to tan pwdr. Strem# 40-1100. http://www.strem.com/catalog/v/40-1100/
Catalog No. 50-901-11577